CO1H324A5CCA CombiPhos Catalysts Inc

2,6-Bis(trifluoromethyl)pyridine-4-boronic acid

  • Cas Number : 1653975-45-1
  • Molecular Weight : 258.92

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart