CO1H324A5CCB CombiPhos Catalysts Inc

2,6-dicyanopyridin-4-ylboronic acid

  • Cas Number : 903513-61-1
  • Molecular Weight : 172.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart