CO1H324A5CD0 CombiPhos Catalysts Inc

2-Methylthiazole-5-boronic acid

  • Cas Number : 1190875-47-8
  • Molecular Weight : 142.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart