CO1H324A5CD6 CombiPhos Catalysts Inc

5-Methyl-2-ethylpyridine-3-boronic acid

  • Cas Number : 2225179-86-0
  • Molecular Weight : 165

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart