CO1H324A5CD8 CombiPhos Catalysts Inc

5-Methyl-2-(cyclopropyl)pyridine-3-boronic acid

  • Cas Number : 2225176-45-2
  • Molecular Weight : 177.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart