CO1H324A5CD9 CombiPhos Catalysts Inc

5-Methyl-2-(tert-butyl)pyridine-3-boronic acid

  • Cas Number : 2225174-37-6
  • Molecular Weight : 193.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart