CO1H324A5CDD CombiPhos Catalysts Inc

5-Bromo-2-(cyclopropyl)pyridine-3-boronic acid

  • Cas Number : 2225175-10-8
  • Molecular Weight : 241.88

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart