CO1H324A5CE0 CombiPhos Catalysts Inc

5-Chloro-2-ethylpyridine-3-boronic acid

  • Cas Number : 2225176-65-6
  • Molecular Weight : 185.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart