CO1H324A5CE3 CombiPhos Catalysts Inc

5-Chloro-2-(tert-butyl)pyridine-3-boronic acid

  • Cas Number : 2225174-11-6
  • Molecular Weight : 213.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart