CO1H324A5CE7 CombiPhos Catalysts Inc

2-Ethyl-5-(cyclopropyl)pyridine-3-boronic acid

  • Cas Number : 2225154-65-2
  • Molecular Weight : 191.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart