CO1H324A5CEE CombiPhos Catalysts Inc

2-Methyl-5-cyanopyridine-3-boronic acid

  • Cas Number : 2225174-22-9
  • Molecular Weight : 161.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart