CO1H324A5CEF CombiPhos Catalysts Inc

2-Ethyl-5-cyanopyridine-3-boronic acid

  • Cas Number : 2225170-93-2
  • Molecular Weight : 175.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart