CO1H324A5CF3 CombiPhos Catalysts Inc

2-Ethyl-5-(methoxycarbonyl)pyridine-3-boronic acid

  • Cas Number : 2225170-95-4
  • Molecular Weight : 209.01

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart