CO1H324A5CF6 CombiPhos Catalysts Inc

2-(tert-Butyl)-5-(trifluoromethyl)pyridine-3-boronic acid

  • Cas Number : 2225179-92-8
  • Molecular Weight : 247.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart