CO1H324A5CF8 CombiPhos Catalysts Inc

5-FLUORO-2-METHYLPYRIDINE-4-BORONIC ACID

  • Cas Number : 1310404-23-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Availability details for
Quantity
  • Availability
  • Add to Cart