CO1H324A5D03 CombiPhos Catalysts Inc

5-METHYL-2-TRIFLUOROMETHYLPYRIDINE-4-BORONIC ACID

  • Cas Number : 2225179-94-0
  • Molecular Weight : 204.94

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart