CO1H324A5D08 CombiPhos Catalysts Inc

5-CYANO-2-TRIFLUOROMETHYLPYRIDINE-4-BORONIC ACID

  • Cas Number : 2225169-26-4
  • Molecular Weight : 215.93

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart