CO1H324A5D0C CombiPhos Catalysts Inc

5-Bromo-2-cyanopyridine-4-boronic acid

  • Cas Number : 2225170-91-0
  • Molecular Weight : 226.82

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart