CO1H324A5D12 CombiPhos Catalysts Inc

5-Chloro-2-(methoxycarbonyl)pyridine-4-boronic acid

  • Cas Number : 2225176-71-4
  • Molecular Weight : 215.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart