CO1H324A5D18 CombiPhos Catalysts Inc

5-Methoxy-2-(methoxycarbonyl)pyridine-4-boronic acid

  • Cas Number : 2225174-46-7
  • Molecular Weight : 210.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart