CO1H324A5D27 CombiPhos Catalysts Inc

4-Fluoro-2-(iso-propoxy)pyridine-5-boronic acid

  • Cas Number : 2225171-59-3
  • Molecular Weight : 198.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart