CO1H324A5D28 CombiPhos Catalysts Inc

4-Fluoro-2-(cyclobutoxy)pyridine-5-boronic acid

  • Cas Number : 2225174-65-0
  • Molecular Weight : 211

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart