CO1H324A5D3C CombiPhos Catalysts Inc

4-Fluoro-2-(thiazol-2-yl)pyridine-5-boronic acid

  • Cas Number : 2225175-79-9
  • Molecular Weight : 224.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart