CO1H324A5D43 CombiPhos Catalysts Inc

(2-ethylpyrimidin-5-yl)boronic acid

  • Cas Number : 2225174-81-0
  • Molecular Weight : 151.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart