CO1H324A5D48 CombiPhos Catalysts Inc

5-Fluoro-2-(2-methylimidazol-1-yl)pyridine-4-boronic acid

  • Cas Number : 2225174-54-7
  • Molecular Weight : 221

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart