CO1H324A5D51 CombiPhos Catalysts Inc

5-Fluoro-2-(oxan-4-yl)pyridine-4-boronic acid

  • Cas Number : 2225171-73-1
  • Molecular Weight : 225.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart