CO1H324A5D53 CombiPhos Catalysts Inc

5-Fluoro-2-(diethylamino)pyridine-4-boronic acid

  • Cas Number : 2225171-88-8
  • Molecular Weight : 212.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart