CO1H324A5D5D CombiPhos Catalysts Inc

5-Fluoro-2-(cyclohexyl)pyridine-4-boronic acid

  • Cas Number : 2225171-74-2
  • Molecular Weight : 223.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart