CO1H324A5D60 CombiPhos Catalysts Inc

2-(Cyclobutyl)pyrimidine-5-boronic acid

  • Cas Number : 1551413-91-2
  • Molecular Weight : 178

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart