CO1H324A5D66 CombiPhos Catalysts Inc

2-Fluoro-6-(n-propyl)pyridine-4-boronic acid

  • Cas Number : 2225154-82-3
  • Molecular Weight : 182.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart