CO1H324A5D6D CombiPhos Catalysts Inc

2-Fluoro-6-(cyclopentyl)pyridine-4-boronic acid

  • Cas Number : 2225180-44-7
  • Molecular Weight : 209.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart