CO1H324A5D77 CombiPhos Catalysts Inc

2-Fluoro-6-bromopyridine-4-boronic acid

  • Cas Number : 2225176-29-2
  • Molecular Weight : 219.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart