CO1H324A5D84 CombiPhos Catalysts Inc

2-Cyclobutyl-6-(piperidin-3-yl)pyridine-4-boronic acid

  • Cas Number : 2225152-45-2
  • Molecular Weight : 260.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart