CO1H324A5D8F CombiPhos Catalysts Inc

(2-(m-tolyl)pyrimidin-5-yl)boronic acid

  • Cas Number : 2225170-06-7
  • Molecular Weight : 214.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart