CO1H324A5DA0 CombiPhos Catalysts Inc

2-Trifluoromethyl-6-(piperidin-4-yl)pyridine-4-boronic acid

  • Cas Number : 2225177-56-8
  • Molecular Weight : 274.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart