CO1H324A5DA9 CombiPhos Catalysts Inc

(2-(4-(trifluoromethyl)phenyl)pyrimidin-5-yl)boronic acid

  • Cas Number : 2225180-48-1
  • Molecular Weight : 268

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart