CO1H324A5DAB CombiPhos Catalysts Inc

(2-(3-methoxyphenyl)pyrimidin-5-yl)boronic acid

  • Cas Number : 2225154-84-5
  • Molecular Weight : 230.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart