CO1H324A5DAE CombiPhos Catalysts Inc

(2-(3-chlorophenyl)pyrimidin-5-yl)boronic acid

  • Cas Number : 2225177-68-2
  • Molecular Weight : 234.45

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart