CO1H324A5DB0 CombiPhos Catalysts Inc

2-(Cyclobutoxy)pyrimidine-5-boronic acid

  • Cas Number : 1646268-09-8
  • Molecular Weight : 194

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart