CO1H324A5DB4 CombiPhos Catalysts Inc

5-(iso-Propyl)-6-trifluoromethylpyridine-3-boronic acid

  • Cas Number : 2225176-68-9
  • Molecular Weight : 233

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart