CO1H324A5DB9 CombiPhos Catalysts Inc

5-Cyclopropyl-6-trifluoromethylpyridine-3-boronic acid

  • Cas Number : 2225154-87-8
  • Molecular Weight : 230.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart