CO1H324A5DC5 CombiPhos Catalysts Inc

2-Bromo-6-iodopyridine-4-boronic acid

  • Cas Number : 2225175-26-6
  • Molecular Weight : 327.71

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart