CO1H324A5DC9 CombiPhos Catalysts Inc

4-(4-Bromopyridin-2-yl)furan-2-boronic acid

  • Cas Number : 2225176-88-3
  • Molecular Weight : 267.87

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart