CO1H324A5DD1 CombiPhos Catalysts Inc

5-(Pyridin-3-yl)thiophene-2-boronic acid

  • Cas Number : 1421762-96-0
  • Molecular Weight : 205.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart