CO1H324A5DD7 CombiPhos Catalysts Inc

2-(Imidazol-1-yl)pyrimidine-5-boronic acid

  • Cas Number : 2225176-97-4
  • Molecular Weight : 189.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart