CO1H324A5DDA CombiPhos Catalysts Inc

5-(Pyrimidin-2-yl)thiophene-2-boronic acid

  • Cas Number : 2225172-37-0
  • Molecular Weight : 206.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart