CO1H324A5DE1 CombiPhos Catalysts Inc

4-(2-Chloropyrimidin-4-yl)furan-2-boronic acid

  • Cas Number : 2225177-86-4
  • Molecular Weight : 224.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart