CO1H324A5DF1 CombiPhos Catalysts Inc

3-(4-Trifluoromethylpyridin-2-yl)phenylboronic acid

  • Cas Number : 2225176-99-6
  • Molecular Weight : 267.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart