CO1H324A5DF3 CombiPhos Catalysts Inc

3-(5-Bromopyridin-2-yl)phenylboronic acid

  • Cas Number : 2225180-53-8
  • Molecular Weight : 277.91

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart