CO1H324A5DF7 CombiPhos Catalysts Inc

3-(4-Chloropyridin-2-yl)phenylboronic acid

  • Cas Number : 2225172-50-7
  • Molecular Weight : 233.46

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 96
Country of Origin US
Availability details for
Quantity
  • Availability
  • Add to Cart